5-Bromo-2-chlorophenol, 98+%
5-Bromo-2-chlorophenol, 98+%
5-Bromo-2-chlorophenol, 98+%
Thermo Scientific Chemicals

5-Bromo-2-chlorophenol, 98+%

CAS: 183802-98-4 | C6H4BrClO | 207.451 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25452.03
also known as B25452-03
Price (USD)
61.65
Online Exclusive
68.60
Save 6.95 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.60
Save 6.95 (10%)
Each
Chemical Identifiers
CAS183802-98-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale orange to pale brown
FormCrystals or powder or crystalline powder or fused solid or chunks
Assay (GC)≥98.0%
Melting Point (clear melt)54.0-60.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only