2-Bromoindene, 98%
2-Bromoindene, 98%
2-Bromoindene, 98%
Thermo Scientific Chemicals

2-Bromoindene, 98%

CAS: 10485-09-3 | C9H7Br | 195.059 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25474.06
also known as B25474-06
Price (USD)
42.65
Special offer
Online exclusive
Ends: 31-Dec-2025
49.70
Save 7.05 (14%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 31-Dec-2025
49.70
Save 7.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS10485-09-3
IUPAC Name2-bromo-1H-indene
Molecular FormulaC9H7Br
InChI KeyCCUYEVNCRQDQRF-UHFFFAOYSA-N
SMILESBrC1=CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
Assay (GC)≥97.5%
FormPowder or fused solid
Melting Point (clear melt)34.0-40.0?C
2-Bromoindene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoindene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. W.E.Billups; John D.Buynak; Dorothy Wolf. Generation of a 1,3-dehydronaphthalene. Evidence for the diyl structure. J. Org. Chem. 1979, 44,(23), 4218-4219.
  2. Dirk Tews; Petra Escarpa Gaede. Synthesis of a novel bridged 2-(cyclopentadienyl)-indene system using Pd-catalyzed Negishi-type cross coupling. Tetrahedron Letters. 2004, 45,(49), 9029-9031.