4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%
4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%
4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%
4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%
Thermo Scientific Chemicals

4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine, 98%

CAS: 13104-56-8 | C22H17N3O | 339.398 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number B25486.02
also known as B25486-02
Price (USD)
64.65
Online Exclusive
72.00
Save 7.35 (10%)
Each
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (USD)
64.65
Online Exclusive
72.00
Save 7.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13104-56-8
IUPAC Name4-(4-methoxyphenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular FormulaC22H17N3O
InChI KeyQAEWAYWLMREGRA-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Purity (DSC)≥97.5%
Melting Point (clear melt)161-163?C
Proton NMRConforms to structure
FormPowder

4'-(4-Methoxyphenyl)-2,2':6',2″-terpyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-(4-Methoxyphenyl)-2,2′:6′,2′′-terpyridine is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Arockiam Anthonysamy; Sengottuvelan Balasubramanian; Vellasamy Shanmugaiah; Narayanasamy Mathivanan. Synthesis, characterization and electrochemistry of 4'-functionalized 2,2':6',2″-terpyridine ruthenium(II) complexes and their biological activity. Dalton Trans. 2008, (16), 2136-2143.
  2. Yu-Zhe Chen; Deng-Hui Wang; Bin Chen; Jian-Ji Zhong; Chen-Ho Tung; Li-Zhu Wu. Visible light-induced synthesis of 3,4-diarylthiophenes from 3,4-diaryl-2,5-dihydrothiophenes. J. Org. Chem. 2012, 77,(16), 6773-6777.