1-Bromo-3,5-dichlorobenzene, 97%
1-Bromo-3,5-dichlorobenzene, 97%
1-Bromo-3,5-dichlorobenzene, 97%
Thermo Scientific Chemicals

1-Bromo-3,5-dichlorobenzene, 97%

CAS: 19752-55-7 | C6H3BrCl2 | 225.894 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25492.06
also known as B25492-06
Price (USD)
22.65
Online Exclusive
24.80
Save 2.15 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
22.65
Online Exclusive
24.80
Save 2.15 (9%)
Each
Chemical Identifiers
CAS19752-55-7
IUPAC Name1-bromo-3,5-dichlorobenzene
Molecular FormulaC6H3BrCl2
InChI KeyDZHFFMWJXJBBRG-UHFFFAOYSA-N
SMILESClC1=CC(Br)=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)71.0-81.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only