2-Phenyl-3-butyn-2-ol, 98%
2-Phenyl-3-butyn-2-ol, 98%
2-Phenyl-3-butyn-2-ol, 98%
Thermo Scientific Chemicals

2-Phenyl-3-butyn-2-ol, 98%

CAS: 127-66-2 | C10H10O | 146.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B25515.06
also known as B25515-06
Price (USD)
45.65
Online exclusive
50.10
Save 4.45
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
45.65
Online exclusive
50.10
Save 4.45
Each
Chemical Identifiers
CAS127-66-2
IUPAC Name2-phenylbut-3-yn-2-ol
Molecular FormulaC10H10O
InChI KeyKSLSOBUAIFEGLT-UHFFFAOYNA-N
SMILESCC(O)(C#C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5% (GC, U.S. Specification)
Identification (FTIR)Conforms (U.S. Specification)
FormCrystals or powder or crystalline powder
CommentPurchased in the U.S. and in other countries
Melting Point (clear melt)47.0-53.0°C (Non-U.S. Specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only