5-Chloro-2-methylphenol, 98%
5-Chloro-2-methylphenol, 98%
5-Chloro-2-methylphenol, 98%
5-Chloro-2-methylphenol, 98%
Thermo Scientific Chemicals

5-Chloro-2-methylphenol, 98%

CAS: 5306-98-9 | C7H7ClO | 142.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25527.14
also known as B25527-14
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
234.65
Online Exclusive
261.00
Save 26.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5306-98-9
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)70.5-76.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only