2-Bromothiophenol, 97%
2-Bromothiophenol, 97%
2-Bromothiophenol, 97%
Thermo Scientific Chemicals

2-Bromothiophenol, 97%

CAS: 6320-02-1 | C6H5ClS | 144.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25537.06
also known as B25537-06
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.90
Save 10.25 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 30-Jun-2026
68.90
Save 10.25 (15%)
Each
Chemical Identifiers
CAS6320-02-1
IUPAC Name2-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyYUQUNWNSQDULTI-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.6320-1.6370 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only