1-Fluoro-3-nitrobenzene, 98+%
1-Fluoro-3-nitrobenzene, 98+%
1-Fluoro-3-nitrobenzene, 98+%
1-Fluoro-3-nitrobenzene, 98+%
Thermo Scientific Chemicals

1-Fluoro-3-nitrobenzene, 98+%

CAS: 402-67-5 | C6H4FNO2 | 141.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B25553.22
also known as B25553-22
Price (USD)
220.65
Online Exclusive
245.00
Save 24.35 (10%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
220.65
Online Exclusive
245.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS402-67-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to green to orange to brown
Refractive Index1.5230-1.5270 @20?C
FormLiquid
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only