Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate
Thermo Scientific Chemicals

Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate

CAS: 7681-38-1 | HNaO4S | 120.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number B25587.0B
also known as B25587-0B
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.20
Save 12.55
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
72.65
Special offer
Online exclusive
Ends: 28-Dec-2026
85.20
Save 12.55
Each
Chemical Identifiers
CAS7681-38-1
IUPAC Namesodium hydrogen sulfate
Molecular FormulaHNaO4S
InChI KeyWBHQBSYUUJJSRZ-UHFFFAOYSA-M
SMILES[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Transparent or white to yellow
FormCrystalline solid or powder or granules
Assay (Aqueous acid-base Titration)≥90.0 to ≤110.0%
pH≤1.2 (5% in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only