Bis(4-chlorophenyl) disulfide, 98+%
Bis(4-chlorophenyl) disulfide, 98+%
Bis(4-chlorophenyl) disulfide, 98+%
Bis(4-chlorophenyl) disulfide, 98+%
Thermo Scientific Chemicals

Bis(4-chlorophenyl) disulfide, 98+%

CAS: 1142-19-4 | C12H8Cl2S2 | 287.216 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25604.06
also known as B25604-06
Price (USD)
62.80
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
62.80
Each
Add to cart
Chemical Identifiers
CAS1142-19-4
IUPAC Name1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene
Molecular FormulaC12H8Cl2S2
InChI KeyZIXXRXGPBFMPFD-UHFFFAOYSA-N
SMILESClC1=CC=C(SSC2=CC=C(Cl)C=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to cream
FormPowder or granules
Assay (GC)≥98.0%
Melting Point (clear melt)69.0-76.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only