3,5-Dimethoxyphenylacetic acid, 98%
Thermo Scientific Chemicals

3,5-Dimethoxyphenylacetic acid, 98%

CAS: 4670-10-4 | C8H14O | 126.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25672.03
also known as B25672-03
Price (USD)
71.90
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
71.90
Each
Chemical Identifiers
CAS4670-10-4
IUPAC Name2-(3,5-dimethoxyphenyl)acetate
Molecular FormulaC10H11O4
InChI KeyFFPAFDDLAGTGPQ-UHFFFAOYSA-M
SMILESCOC1=CC(OC)=CC(CC([O-])=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Appearance (Color)White to cream or pale yellow
Melting Point (clear melt)101-106?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only