2,6-Diacetylpyridine, 99%
2,6-Diacetylpyridine, 99%
2,6-Diacetylpyridine, 99%
Thermo Scientific Chemicals

2,6-Diacetylpyridine, 99%

CAS: 1129-30-2 | C9H9NO2 | 163.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B25678.06
also known as B25678-06
Price (USD)
157.65
Online Exclusive
175.00
Save 17.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
157.65
Online Exclusive
175.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS1129-30-2
IUPAC Name1-(6-acetylpyridin-2-yl)ethan-1-one
Molecular FormulaC9H9NO2
InChI KeyBEZVGIHGZPLGBL-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC(=N1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)≥98.5%
Melting Point (clear melt)77.0-84.0°C
FormCrystals or powder or crystalline powder or lumps
Identification (FTIR)Conforms
2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diacetylpyridine is used as intermediate in organic synthesis. It is a precursor to ligands in coordination chemistry.

Solubility
Soluble in water, chloroform, and DMSO.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. E. C. Alyea & P. H. Merrell. Terdentate NNN Donor Ligands Derived from 2,6-Diacetylpyridine. Synthesis and Reactivity in Inorganic and Metal-Organic Chemistry. 1974, 4 (6), 535-544.
  2. Wanda Radecka-Paryzek. The synthesis and characterization of the macrocyclic and ring-opened complexes formed in the reaction of the lanthanides with 2,6-diacetylpyridine and hydrazine. Inorganica Chimica Acta. 1981, 52 261-268.