2,5-Hexanedione, 97%
2,5-Hexanedione, 97%
2,5-Hexanedione, 97%
Thermo Scientific Chemicals

2,5-Hexanedione, 97%

CAS: 110-13-4 | C6H10O2 | 114.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B25686.14
also known as B25686-14
Price (USD)
37.65
Online Exclusive
41.40
Save 3.75 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
37.65
Online Exclusive
41.40
Save 3.75 (9%)
Each
Chemical Identifiers
CAS110-13-4
IUPAC Namehexane-2,5-dione
Molecular FormulaC6H10O2
InChI KeyOJVAMHKKJGICOG-UHFFFAOYSA-N
SMILESCC(=O)CCC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to yellow to orange to brown
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4230-1.4280 @ 20?C
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

Solubility
Miscible with alcohol, water and diethyl ether.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and strong reducing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Primary amines can be protected by conversion to 2,5-dimethylpyrroles, from which they can be released by reaction with hydroxylamine hydrochloride: J. Chem. Soc., Perkin 1, 2801 (1984). The method has been found useful for protecting amino sugars in oligosaccharide synthesis: J. Org. Chem., 63, 4570 (1998), and also in the protection of an arylamine during Cu(I) promoted methoxylation of a ring bromo or iodo substituent; Synthesis, 1599 (1998).
  2. Sacia, E. R.; Deaner, M. H.; Louie, Y. L.; Bell, A. T. Synthesis of biomass-derived methylcyclopentane as a gasoline additivevia aldol condensation/hydrodeoxygenation of 2,5-hexanedione. Green Chem. 2015, 17 (4), 2393-2397.
  3. Chambon, F.; Rataboul, F.; Pinel, C.; Cabiac, A.; Guillon, E.; Essayem, N. Conversion of cellulose to 2,5-hexanedione using tungstated zirconia in hydrogen atmosphere. Appl. Catal., A 2015, 504, 664-671.