2',6'-Dimethylacetanilide, 97%
2',6'-Dimethylacetanilide, 97%
2',6'-Dimethylacetanilide, 97%
Thermo Scientific Chemicals

2',6'-Dimethylacetanilide, 97%

CAS: 2198-53-0 | C10H13NO | 163.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B25687.22
also known as B25687-22
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS2198-53-0
IUPAC NameN-(2,6-dimethylphenyl)acetamide
Molecular FormulaC10H13NO
InChI KeyNRPTXWYBRKRZES-UHFFFAOYSA-N
SMILESCC(=O)NC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or lumps
Assay (GC)≥96.0%
Melting Point (clear melt)178.0-184.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only