beta-Cyclocitral, 90+%
beta-Cyclocitral, 90+%
beta-Cyclocitral, 90+%
Thermo Scientific Chemicals

beta-Cyclocitral, 90+%

CAS: 432-25-7 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B25700.18
also known as B25700-18
Price (USD)
697.65
Special offer
Online exclusive
Ends: 30-Jun-2026
820.00
Save 122.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
697.65
Special offer
Online exclusive
Ends: 30-Jun-2026
820.00
Save 122.35 (15%)
Each
Chemical Identifiers
CAS432-25-7
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥90.0%
Refractive Index1.4850-1.5050 @ 20?C
Free acid (titration)≤3.0%
View more
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.

Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol. Insoluble in water.

Notes
Air sensitive. Store away from air, oxidizing agents.
RUO – Research Use Only

General References:

  1. Volker Buss; Klaus Kolster; Ute Wingen; Lothar Simon. Chiral recognition of the twisted cyclohexene conformation in .beta.-cyclocitral Schiff bases. J. Am. Chem. Soc. 1984, 106 (16), 4621-4622.
  2. Michael Klein; Ute Wingen; Volker Buss. Chiral .beta.-cyclocitral Schiff base: a combined spectroscopic and theoretical approach to a twisted enimine structure. J. Am. Chem. Soc. 1987, 109 (21), 6486-6492.