Ethyl 2,4-dioxo-5-methylhexanoate, 95%
Ethyl 2,4-dioxo-5-methylhexanoate, 95%
Ethyl 2,4-dioxo-5-methylhexanoate, 95%
Ethyl 2,4-dioxo-5-methylhexanoate, 95%
Thermo Scientific Chemicals

Ethyl 2,4-dioxo-5-methylhexanoate, 95%

CAS: 64195-85-3 | C9H14O4 | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number B25735.14
also known as B25735-14
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
156.65
Special offer
Online exclusive
Ends: 30-Jun-2026
184.00
Save 27.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS64195-85-3
IUPAC Nameethyl 5-methyl-2,4-dioxohexanoate
Molecular FormulaC9H14O4
InChI KeyHBJPUWYQVWIKSD-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)CC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange
FormLiquid
Assay (GC)≥94.0%
Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2,4-dioxo-5-methylhexanoate is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only
Issa Yavari; Mohammad Bayat. Synthesis of Highly Functionalized Cyclobutene Derivatives. Monatshefte für Chemie / Chemical Monthly. 2003, 134 (9), 1221-1227.