Buffers for RNA Transctription and Post-Transcriptional Modifications
Thermo Scientific™

Buffers for RNA Transctription and Post-Transcriptional Modifications

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityChemical Name or MaterialConcentration
B3277AF200 μLT4 RNA Ligase 2 Reaction Buffer10X
F8156AF200 μLTranscription Buffer10X
B23AF800 μLCapping Buffer10X
B2348AF1000 μLPoly(A) Polymerase Buffer10X
B1299AF200 μLT4 RNA Ligase Reaction Buffer10X
Catalog number B3277AF
Price (USD)
58.00
Each
Quantity:
200 μL
Chemical Name or Material:
T4 RNA Ligase 2 Reaction Buffer
Concentration:
10X
Price (USD)
58.00
Each

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialT4 RNA Ligase 2 Reaction Buffer
IncludesT4 RNA Ligase 2 Reaction Buffer, 200 μL

Store at -20°C.
Concentration10X
For Use With (Application)RNA Ligation
Physical FormLiquid
Quantity200 μL
Unit SizeEach