Tyramide Conjugates
Invitrogen™

Tyramide Conjugates

Thermo Fisher Scientific offers a wide variety of bright Alexa Fluor™, Alexa Fluor™ Plus or Biotin XX tyramide conjugates. These tyramide conjugates are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConjugateExcitation/Emission
B40953Alexa Fluor™ 488495/519 nm
B40952Alexa Fluor™ 350347/442 nm
B40954Alexa Fluor™ 546556/573 nm
B40955Alexa Fluor™ 555555/565 nm
B40956Alexa Fluor™ 568579/604 nm
B40957Alexa Fluor™ 594591/617 nm
B40958Alexa Fluor™ 647650/668 nm
B56131Alexa Fluor™ Plus 750750/790 nm
B40951Biotin-XX
Catalog number B40953
Price (USD)
534.64
Special offer
Online exclusive
Ends: 31-Dec-2026
652.00
Save 117.36
Each
Conjugate:
Alexa Fluor™ 488
Excitation/Emission:
495/519 nm
Price (USD)
534.64
Special offer
Online exclusive
Ends: 31-Dec-2026
652.00
Save 117.36
Each
Thermo Fisher Scientific offers a wide variety of bright Alexa Fluor™, Alexa Fluor™ Plus or Biotin XX tyramide conjugates. These tyramide conjugates are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD). TSA is used to amplify signals up to 200 fold versus standard imaging methods. This reagent can be used with any antibody conjugated to HRP for tyramide signal amplification to detect low abundance targets. Up to seven-plex is possible in multiplexable fluorescent imaging.

These tyramide reagents are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD). TSA is used to amplify signal up to 200-fold versus standard imaging methods. These reagents are provided stand-alone, but are also available as part of our SuperBoost™ tyramide signal amplification kits (available separately). They can be used with any antibody conjugated to HRP for tyramide signal amplification for detection of low abundance targets. Up to seven-plex is possible in multiplex fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).

Features include:
• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with TO-PRO-3, secondary antibodies, and other SuperBoost kits
• Requires 10–100 times less primary antibody then standard ICC/IHC/ISH experiments

Tyramide reagents, when used as directed, can increase sensitivity up to 200 times over standard imaging methods. These reagents help sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal. They are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a tyramide reagent can be imaged using any type of microscope, producing high-resolution multiplex images.

Dissolve the Alexa Fluor™ or Biotin-XX tyramide reagent in 150 μL of DMSO to prepare 100X stock. Tyramide may coat the sides of the vial so invert the vial several times to ensure all reagent is dissolved in solution.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateAlexa Fluor™ 488
Detection MethodFluorescence
Excitation/Emission495/519 nm
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
ClonalityNone
For Use With (Application)Immunohistochemistry (IHC), Immunofluorescence (IF), Immunocytochemistry (ICC), ISH
Product LineAlexa Fluor
Quantity1 vial, 150 slides
Unit SizeEach
Contents & Storage
• 1 vial Alexa Fluor Tyramide Reagent sufficient for 150 slides

Store at 2°C to 8°C. When stored as directed, the product is stable for 6 months after receiving.

Citations & References (2)

Citations & References
Abstract
Sensitive immunofluorescent staining of cells via generation of fluorescent nanoscale polymer films in response to biorecognition.
Authors:Avens HJ, Berron BJ, May AM, Voigt KR, Seedorf GJ, Balasubramaniam V, Bowman CN
Journal:J Histochem Cytochem
PubMed ID:21339175
Immunofluorescent staining is central to nearly all cell-based research, yet only a few fluorescent signal amplification approaches for cell staining exist, each with distinct limitations. Here, the authors present a novel, fluorescent polymerization-based amplification (FPBA) method that is shown to enable similar signal intensities as the highly sensitive, enzyme-based tyramide ... More
Immunohistochemical Identification of Human Skeletal Muscle Macrophages.
Authors:Kosmac K, Peck BD, Walton RG, Mula J, Kern PA, Bamman MM, Dennis RA, Jacobs CA, Lattermann C, Johnson DL, Peterson CA,
Journal:Bio Protoc
PubMed ID:30148186
Macrophages have well-characterized roles in skeletal muscle repair and regeneration. Relatively little is known regarding the role of resident macrophages in skeletal muscle homeostasis, extracellular matrix remodeling, growth, metabolism and adaptation to various stimuli including exercise and training. Despite speculation into macrophage contributions during these processes, studies characterizing macrophages in ... More