SuperBoost™ Goat anti-Rabbit Poly HRP
Invitrogen™
SuperBoost™ Goat anti-Rabbit Poly HRP
SuperBoost™ Goat anti-Rabbit Poly HRP is a component in SuperBoost™ tyramide signal amplification kits made available separately here. SuperBoost™ GoatRead more
Have Questions?
Catalog NumberQuantity
B4096222.5 mL Bottle (For 150 slides)
Catalog number B40962
Price (USD)
396.00
Each
Quantity:
22.5 mL Bottle (For 150 slides)
Price (USD)
396.00
Each
SuperBoost™ Goat anti-Rabbit Poly HRP is a component in SuperBoost™ tyramide signal amplification kits made available separately here. SuperBoost™ Goat anti-Rabbit Poly HRP can be used with any HRP sustrate such as tyramide, DAB for signal amplification and detection. The molar enzyme/antibody protein ratio has an average value of '4'. This reagent is provided at a ready-to-use 1X concentration, enough for 150 slides.
When used as directed, this reagent with other SuperBoost reagents can be used for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH) with up to 200 times more sensitivity than standard methods. For standout research, SuperBoost reagents sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal. SuperBoost reagents are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a SuperBoost reagent can be imaged using any type of microscope, producing high-resolution multiplex images.
Features of the SuperBoost reagents include:
• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with TO-PRO™-3, secondary antibodies, and other SuperBoost kits
• Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineSuperBoost
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
ConjugateHRP
Quantity22.5 mL Bottle (For 150 slides)
Unit SizeEach
Contents & Storage
22.5 mL at 1X concentration in dropper bottle
Store at 2-8°C. When stored as directed, the product is stable for 6 months after receiving.