Cindion™ Adsorption Module
Cindion™ Adsorption Module
Cindion™ Adsorption Module
Cindion™ Adsorption Module
Thermo Scientific™
Cindion™ Adsorption Module
Determine adsorbable organic halogens (AOX) or adsorbable organic fluorine (AOF) by adding the Thermo Scientific™ Cindion™ Adsorption Module to the Thermo Scientific™ Cindion™ Combustion Ion Chromatography System.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51006430Cindion Combustion IC System
Catalog number B51006430
Price (USD)
-
For Use With (Equipment):
Cindion Combustion IC System
Add the Cindion Adsorption Module to the Cindion Combustion IC (C-IC) System to measure AOX or AOF by C-IC. This adsorption module enables the use of granular activated carbon (GAC) columns to screen aqueous samples for the potential presence of toxic compounds. This includes screening for per- and polyfluoroalkyl substances (PFAS) using US EPA Method 1621.
Pair the Cindion Adsorption Module with other key components of the Cindion C-IC System to screen for PFAS using US EPA Method 1621.
Specifications
Capacity6 Samples
Dimensions (D x W x H)49 cm x 46 cm 56 cm
For Use With (Application)AOX, AOF (PFAS Screening per US EPA Method 1621) for Aqueous Samples
For Use With (Equipment)Cindion Combustion IC System
Weight (English)36.8 lb
Weight (Metric)16.7 kg
TypeAdsorption Module
Unit SizeEach
Frequently asked questions (FAQs)
Can the Cindion C-IC system be used for US EPA Method 1621 (Adsorbable Organic Fluorine (AOF) by C-IC)?
Yes. The Cindion C-IC system supports US EPA Method 1621-compliant AOF analysis using a fully PFAS-free workflow that features an adsorption module that can process up to six samples in parallel using granular activated carbon (GAC) columns.