Cindion™ Blunt-Point Needles
Thermo Scientific™
Cindion™ Blunt-Point Needles
Pair these Thermo Scientific™ Cindion™ Blunt-Point Needles with Thermo Scientific™ Cindion™ PFAS-Free Syringes to withdraw and dispense liquid samples for combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
B51006442250 or 500 μL
B51006441Up to 100 μL syringes
Catalog number B51006442
Price (CLP)
-
Volume (Metric):
250 or 500 μL
Withdraw and dispense liquid samples for combustion-ion chromatography (C-IC) analysis using the Cindion C-IC System with the Cindion Blunt-Point Needles. These blunt-point needles attach to the Cindion PFAS-Free Syringes to dispense samples into the liquids boat prior to C-IC analysis.
These needles are used for the liquid sample configuration of the Cindion Combustion IC System.
  • Pack of 6
  • Available in 2 options: Needles for up to 100 µL syringes and needles for 250 or 500 µL syringes
  • Use with Cindion PFAS-Free Syringes
Specifications
Dimensions (D x W x H)1 cm x 10 cm x 1 cm
For Use With (Application)Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System
MaterialStainless Steel
Quantity6
Volume (Metric)250 or 500 μL
TypeNeedle
Unit SizePack of 6