Cindion™ Boat-Guiding Tube
Thermo Scientific™
Cindion™ Boat-Guiding Tube
Ensure smooth delivery of the sample boat to the combustion oven for combustion ion chromatography analysis with the Thermo Scientific™ Cindion™ Boat-Guiding Tube.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51006444Cindion Combustion IC System
Catalog number B51006444
Price (USD)
-
For Use With (Equipment):
Cindion Combustion IC System
Use this boat-guiding tube to ensure smooth delivery of the sample boat to the combustion oven in the Thermo Scientific™ Cindion™ Combustion IC (C-IC) System. This glass tube surrounds the sample boat to protect the sample prior to C-IC analysis.
  • This boat-guiding tube can be used with the solid- and liquid-sample configurations of the Cindion Combustion IC System.
Specifications
Dimensions (D x W x H)2 cm x 16 cm x 2 cm
For Use With (Application)Combustion Ion Chromatography
For Use With (Equipment)Cindion Combustion IC System
MaterialBorosilicate Glass
TypeCombustion/Absorption Module Accessory
Unit SizeEach