Cindion™ Quartz Wool
Thermo Scientific™
Cindion™ Quartz Wool
Thermo Scientific™ Cindion™ Quartz Wool helps facilitate combustion of liquid samples during combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion Ion Chromatography System.
Have Questions?
Catalog NumberWeight (Metric)
B5100645110 g
Catalog number B51006451
Price (MXN)
-
Weight (Metric):
10 g
Use this quartz wool to facilitate combustion of liquid samples during combustion ion chromatography (C-IC) analysis. Liquid samples are injected into the quartz wool contained within the liquids sample boat.
This quartz wool is used in the liquid-sample configuration of the Cindion Combustion IC System.
Specifications
For Use With (Application)Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System
MaterialQuartz Wool
Weight (Metric)10 g
Unit SizeEach