Cindion™ Carbon Columns
Thermo Scientific™
Cindion™ Carbon Columns
Thermo Scientific™ Cindion™ Carbon Columns are used for sample adsorption with the Thermo Scientific™ Cindion™ Adsorption Module prior to combustion ion chromatography analysis with the Thermo Scientific™ Cindion™ Combustion Ion Chromatography System.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51006452Cindion Combustion IC System
Catalog number B51006452
Price (USD)
-
For Use With (Equipment):
Cindion Combustion IC System
Perform sample adsorption with the Cindion Adsorption Module for combustion ion chromatography (C-IC) analysis using the Cindion Combustion IC System with Cindion Carbon Columns. These columns contain granular activated carbon to measure adsorabable organic halogens (AOX) and adsorbable organic fluorine (AOF) by C-IC.
These carbon columns are used in the solid-sample configuration of the Cindion Combustion IC System that is compatible with US EPA Method 1621. This configuration can be used to:
  • Measure AOX
  • Screen for PFAS by measuring AOF by C-IC following US EPA Method 1621
Specifications
Dimensions (D x W x H)12 cm x 6 cm x 14 cm
For Use With (Application)Combustion Ion Chromatography, AOF by CIC, AOX by CIC
For Use With (Equipment)Cindion Combustion IC System
MaterialGranular Activated Carbon
Quantity100
TypeCarbon Column
Unit SizePack of 100