Cindion™ Column Holder
Cindion™ Column Holder
Cindion™ Column Holder
Thermo Scientific™
Cindion™ Column Holder
Pair Thermo Scientific™ Cindion™ Column Holder with Thermo Scientific™ Cindion™ Carbon Columns to perform sample adsorption using the Thermo Scientific™ Cindion™ Adsorption Module prior to combustion IC analysis with the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51006453Cindion Combustion IC System
Catalog number B51006453
Price (USD)
-
For Use With (Equipment):
Cindion Combustion IC System
Perform sample adsorption for combustion ion chromatography (C-IC) analysis using the Cindion Adsorption Module. The Cindion Column Holder holds the Cindion Carbon Columns for sample adsorption during analysis.
This column holder is used in the solid-sample configuration of the Cindion Combustion IC System that is compatible with US EPA Method 1621. This configuration can be used to:
  • Measure AOX
  • Screen for PFAS by measuring AOF by C-IC following US EPA Method 1621
Specifications
Dimensions (D x W x H)1 cm x 20 cm x 1 cm
For Use With (Application)Combustion Ion Chromatography, AOX by CIC, AOF by CIC
For Use With (Equipment)Cindion Combustion IC System
MaterialPolycarbonate
TypeCarbon Column Holder
Unit SizeEach