TriPlus™ RSH SMART EFOX Sample Prep Station
Thermo Scientific™
TriPlus™ RSH SMART EFOX Sample Prep Station
Automate dispersive liquid-liquid microextraction (DLLME) sample preparation with the Thermo Scientific™ TriPlus™ RSH SMART EFOX Sample Preparation Station, a flexible platform that helps laboratories increase throughput, reduce manual handling, and improve workflow consistency.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51009973TriPlus RSH SMART
Catalog number B51009973
Price (USD)
-
For Use With (Equipment):
TriPlus RSH SMART
Increase laboratory productivity with the Thermo Scientific TriPlus RSH SMART EFOX Sample Preparation Station for automated dispersive liquid-liquid microextraction (DLLME) workflows. The TriPlus RSH SMART EFOX Sample Prep Station reduces manual handling, improves reproducibility, and supports high-throughput sample preparation with automated liquid handling, extraction, concentration, and clean-up steps. Designed for EFOX workflows, the platform helps laboratories streamline operations and deliver consistent analytical results.
Simplify complex sample preparation
  • Coordinate multiple preparation steps (sample extraction, pre-concentration and clean-up) within a single automated sequence
  • Standardize workflow execution across operators and laboratory sites
  • Minimize manual touchpoints throughout the preparation process
  • Reduce solvent consumption compared to traditional extraction approaches
  • Simplify preparation of liquid and solid food samples, and environmental testing samples
Adapt to evolving laboratory needs
  • Minimize variability associated with manual procedures
  • Improve laboratory capacity through automated preparation of larger sample batches
  • Support multiple sample preparation protocols on a single platform
  • Configure modules and accessories to meet application-specific requirements
  • Integrate seamlessly with EFOX end-to-end solutions for contaminant testing
Maximize walkaway operation
  • Enable unattended processing of sample batches
  • Automate liquid handling, reagent addition, extraction, and sample transfer steps
  • Manage samples, reagents, and consumables through automated handling routines
Streamline workflow integration
  • Connect with Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS)
  • Support end-to-end EFOX workflow implementation from sample preparation through analysis
  • Enable automated dispersive liquid-liquid microextraction (DLLME) within a standardized workflow
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only - Not For Diagnostic Procedures.
Specifications
For Use With (Application)Environmental, Food Safety Testing
ModelCompatible with EFOX instrument platforms
QuantityEach
SoftwareChromeleon CDS software
Tool ExchangeAutomatic Tool Change (ATC)
For Use With (Equipment)TriPlus RSH SMART
Product TypeStandalone Extended Rail
Sample Capacity108 x 2 mL, 15 x 10 mL, 60 x 20 mL Vials
Unit SizeEach
Frequently asked questions (FAQs)
How can I determine if the EFOX workflow meets my performance requirements?
Thermo Fisher Scientific application specialists can review your application and performance objectives to help provide guidance on whether the EFOX solution is appropriate for your workflow. Product specifications, application data, and demonstrations may be available to support your evaluation. Your lab will determine suitability and validating performance under their actual operating conditions.
What EFOX workflow implementation support is available?
Thermo Fisher Scientific representatives support EFOX implementation with instrument installation, workflow setup, application onboarding, on-site training, and ongoing technical support to help you get up and running.
Can the EFOX workflow be customized for my application?
Thermo Fisher Scientific application specialists can help provide configuration guidance and implementation support for the EFOX workflow based on your specific application and throughput requirements. Your lab will determine suitability and validating performance under their actual operating conditions.
How does the Thermo Scientific TriPlus RSH SMART EFOX Sample Prep Station integrate into the EFOX workflow?
The Thermo Scientific TriPlus RSH SMART EFOX Sample Prep Station is designed as part of the end-to-end EFOX solution, with automated sample separation, liquid chromatography systems, mass spectrometry, and software components. Your Thermo Fisher Scientific representative can help provide configuration guidance and implementation support that aligns with your laboratory's process and requirements.