PCR Plate, 384-well, barcoded
Thermo Scientific™

PCR Plate, 384-well, barcoded

Reduce well-to-well variability with 384-Well PCR Plate Full Skirted PCR Plates, thin walled for efficient heat transfer and compatible with most major thermal cyclers.
Have Questions?
Catalog NumberQuantity
BC138450 plates
Catalog number BC1384
Quantity:
50 plates

384-Well Full Skirted PCR Plates are thin walled for efficient heat transfer and compatible with most major thermal cyclers.

The Standard Thermo Scientific 384-well plate, Barcoded, is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40 μL and a working volume of 25 μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.

Highlights of the Standard Thermo Scientific 384-well plate

  • Fully skirted for use with automated systems
  • Compatible with all leading 384 block thermal cyclers
  • 40 μL maximum well volume, 25 μL working volume
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are available and optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection (BC1384W)
  • Cut corner: A24

Applications

  • PCR
  • qPCR/Real-Time PCR
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging50 Plate
Product TypePCR Microplate
Quantity50 plates
Reaction SpeedStandard
Skirt StyleSkirted
Volume (Metric) Working25 μL
BarcodeIncludes Barcode
ColorNatural
MaterialPolypropylene
No. of Wells384
Unit SizeEach

Frequently asked questions (FAQs)

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.