Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
SuperPlate PCR Plate, 96-well, semi-skirted, flat deck, white, barcoded
Thermo Scientific™

SuperPlate PCR Plate, 96-well, semi-skirted, flat deck, white, barcoded

The Thermo Scientific 96-Well Semi-Skirted Plates, Flat Deck, Barcoded and White, are compatible with all ABI platforms, including sequencers, withRead more
Have Questions?
Catalog NumberQuantity
BC2400W25 plates
Catalog number BC2400W
Price (USD)
274.00
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
274.00
Each
The Thermo Scientific 96-Well Semi-Skirted Plates, Flat Deck, Barcoded and White, are compatible with all ABI platforms, including sequencers, with no adapters necessary. The industry-leading manufacturing process and innovative product design deliver reliable, high-performance products that generate accurate and reproducible PCR data.

The SuperPlate versions are 4x more rigid than standard plates, making these plates ideal for robotic handling. They feature a cut corner at the A12 location.

Highlights

• Directly compatible with all major major thermal cyclers, including ABI platforms including sequencers with no adapters necessary
• Flat plate deck facilitates sealing and handling
• The SuperPlate version provides 4x more rigidity for superior robotic handling
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• Black alpha-numeric lettering allows for easy well identification
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A12

Applications

• qPCR/Real-Time PCR

Barcoding
The barcodes are the industry standard code 128. The label itself can withstand temperatures from -196°C up to 120°C to ensure that the barcodes stay on the plate throughout your entire workflow. It also has a special coating to virtually eliminate any ‘rubbing off' of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.

Related Products
PCR Plate, 96-well, semi-skirted, flat deck, barcoded
PCR Plate, 96-well, semi-skirted, flat deck, white, barcoded
SuperPlate PCR Plate, 96-well, semi-skirted, flat deck, barcoded
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Working0.2 mL
BarcodeIncludes Barcode
ColorWhite
MaterialPolypropylene
No. of Wells96
Product LineSuperPlate
Unit SizeEach