Sterilin™ Pipette Tips
Actual product may vary
Thermo Scientific™

Sterilin™ Pipette Tips

Rely on Thermo Scientific™ Sterilin™ Pipette Tips for excellent sample visibility and minimal fluid retention with your Gilson, Eppendorf and universal-style pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorTip Style
BCT205 to 10 μLClearEppendorf
BCT255 to 200 μLYellowGilson
BCT305 to 200 μLYellowUniversal
BCT70100 to 1000 μLBlueUniversal
BCT1001000 to 5000 μLClearGilson
Catalog number BCT20
Price (USD)
-
Volume (Metric):
5 to 10 μL
Color:
Clear
Tip Style:
Eppendorf
Rely on Thermo Scientific™ Sterilin™ Pipette Tips for excellent sample visibility and minimal fluid retention with your Gilson, Eppendorf and universal-style pipettes.
  • Manufactured from virgin polypropylene and metal-free pigments
  • High translucency for excellent visibility of sample
  • Hydrophobic surface to minimize fluid retention
  • Precision molded for clean delivery
  • Supplied nonsterile and autoclavable at 121°C
  • Supplied bulk packed, an economical choice for the busy laboratory

Ordering Information

  • Not available in all geographic regions
Specifications
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
For Use With (Equipment)Eppendorf
FormatBulk
Product LineSterilin
SterilityNon-sterile
Tip StyleEppendorf
Volume (Metric)5 to 10 μL
Unit SizeEach