Product Image
Thermo Scientific™
Maximum Recovery Diluent Ready Bag
Maintain viability and maximize the recovery of microorganisms with a protective and isotonic diluent in convenient ready prepared bags.
Have Questions?
Catalog number BM0204T
Price (MXN)
-
Maintain viability and maximize the recovery of microorganisms with protective and isotonic Thermo Scientific™ Oxoid™ Maximum Recovery Diluent Ready Bag. Maximum Recovery Diluent Ready Bags are designed for use in food testing, by laboratories with large numbers of samples, requiring large volumes of broth or diluent.
Shorten your workflow with easy-to-use Maximum Recovery Diluent Ready Bag. Ready to go when you are, the bags are highly adaptable to laboratory workflows and automated equipment, such as dilutors, with no further supplementation required. 
  • Cost-effective: no need for weighing of media or sterilization, reducing the burden of media preparation, which frees up resources and equipment.
  • Easy to use: easy to dispense with minimum training
  • Safe: eliminate handling of glassware, weighing of culture media, and the need to add supplementary reagents. Empty bags can be disposed of with normal laboratory waste
For added convenience, it is recommended that Ready Bag Enrichment Media be dispensed using the Thermo Scientific Automated Diluter (DB4150A).
Specifications
ColorColorless, clear
DescriptionMaximum Recovery Diluent Ready Bag
Fill Weight3020mL ± 20mL
Format3L bag with connection ports in a box
Quantity4/Pk.
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
2 to 25°C