Dry-Bags™ Tubing Connector
Dry-Bags™ Tubing Connector
Dry-Bags™ Tubing Connector
Dry-Bags™ Tubing Connector
Thermo Scientific™
Dry-Bags™ Tubing Connector
Use this stainless steel connector to attach Thermo Scientific™ peristaltic pump tubing to the filling tubing that is part of each Thermo Scientific™ Dry-Bag™ .
Have Questions?
Catalog NumberPort Size
BM9901A77mm
Catalog number BM9901A
Price (TWD)
-
Port Size:
77mm
Attach Thermo Scientific™ Pump Tubing to Thermo Scientific™ Dry-Bags™ tubing using a durable, stainless steel Thermo Scientific Dry-Bags Tubing Connector.
This product is for use with pump units DB2000A and DB3000A.
Contact us to arrange a demonstration of the Peristaltic Pump and its accessories.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionDry-Bags Tubing Connector
For Use WithDry-Bags
IncludesIncludes tubing connector only
Port Size77mm
Diameter (English) Inner3.03 in.
Diameter (Metric) Inner77 mm
MaterialStainless Steel
Product TypeAccessories
Unit SizeEach