Product Image
Invitrogen™

eBioscience™ Annexin V Apoptosis Detection Kits

Kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityExcitation/EmissionConjugate
BMS500BT-20
also known as BMS500BT/20
20 TestsNA, 535/NA, 617Biotin, Propidium Iodide
88-8005-74200 Tests488, 535/520, 617FITC, Propidium Iodide
88-8006-7250 Tests405, 546/450, 647eFluor 450, 7-AAD
88-8006-74200 Tests405, 546/450, 647eFluor 450, 7-AAD
88-8007-74200 Tests650, 535/660, 617APC, Propidium Iodide
88-8008-7250 TestsPCP-eFluor 710
88-8008-74200 TestPCP-eFluor 710
BMS500FI-100100 Tests488, 535/520, 617FITC, Propidium Iodide
BMS500FI-2020 Tests488, 535/520, 617FITC, Propidium Iodide
BMS500FI-300300 Tests488, 535/520, 617FITC, Propidium Iodide
88-8102-7250 TestsPE, 7-AAD
88-8102-74200 TestsPE, 7-AAD
88-8103-7250 TestsPE/Cy7
88-8103-74200 TestsPE/Cy7
BMS500BT-100
also known as BMS500BT/100
100 TestsNA, 535/NA, 617Biotin, Propidium Iodide
BMS500BT-300
also known as BMS500BT/300
300 TestsNA, 535/NA, 617Biotin, Propidium Iodide
88-8005-7250 Tests488, 535/520, 617FITC, Propidium Iodide
88-8007-7250 Tests650, 535/660, 617APC, Propidium Iodide
Catalog number BMS500BT-20
also known as BMS500BT/20
Price (USD)
168.65
Online exclusive
181.00
Save 12.35
Each
Quantity:
20 Tests
Excitation/Emission:
NA, 535/NA, 617
Conjugate:
Biotin, Propidium Iodide
Price (USD)
168.65
Online exclusive
181.00
Save 12.35
Each
Easily distinguish cells undergoing early and late apoptosis with eBioscience and Annexin V Apoptosis Detection kits for flow cytometry analysis. These kits utilize viability dye-conjugated annexin V to label phosphatidylserine (PS), an early marker of apoptosis, on the extracellular membrane. In late-stage apoptosis, as cell integrity is lost, both annexin V and the viability dye traverse and bind PS on the interior of the plasma membrane.
Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or fixable viability dyes such as eFluor 450. These cells will stain with annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late-stage apoptosis, the cell membrane loses integrity, thereby allowing annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (annexin V, viability dye-positive) from the early-stage apoptotic cells (annexin V positive, viability dye-negative).

Current viability stains available with the eBioscience Annexin V Apoptosis Detection kits include FITC, eFluor 450, allophycocyanin (APC), phycoerythrin (PE), PE-cyanine7, and biotin.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptioneBioscience Annexin V-Biotin Apoptosis Detection Kit
Excitation/EmissionNA, 535/NA, 617
For Use With (Equipment)Flow Cytometer
Product TypeApoptosis Detection Kit
Quantity20 Tests
Shipping ConditionWet Ice
ConjugateBiotin, Propidium Iodide
Unit SizeEach
Contents & Storage
Contains:
• Binding Buffer (4X), 1 bottle (50 mL)
• rh Annexin V-Biotin, 1 vial (0.1 mL)
• Propidium Iodide (20 μg/mL), 1 vial (1.8 mL). Handling Conditions:
Store at 2-8°C. Avoid temperature fluctuations.

Frequently asked questions (FAQs)

Can I use eFluor 450-conjugated antibodies with eBioscience Annexin V Apoptosis Detection kits?

Yes, eFluor 450-conjugated antibodies are compatible with eBioscience Annexin V Apoptosis Detection kits.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.