Product Image
Invitrogen™

eBioscience™ Propidium Iodide

Propidium Iodide (PI) is a standard reagent used for assessing cell viability and exclusion of non-viable cells in flow cytometry.Read more
Have Questions?
Catalog NumberQuantity
BMS500PI1.5 mL
Catalog number BMS500PI
Price (USD)
111.00
Each
Quantity:
1.5 mL
Price (USD)
111.00
Each
Propidium Iodide (PI) is a standard reagent used for assessing cell viability and exclusion of non-viable cells in flow cytometry. PI binds to double stranded DNA, but is excluded from cells with intact plasma membranes. PI should be analyzed in the PE channel when used as a counterstain for Annexin V FITC. 7-AAD can be used in place of PI when using Annexin V PE. The advantage of 7-AAD over PI is that there is minimal spectral overlap between these emissions. Choose propidium Iodide as a counterstain with Annexin V apoptosis assays.

Reactivity/Species
Human, Mouse, Rat

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Dye TypeCell-Impermeant
Emission Wavelength535/617
Quantity1.5 mL
ColorRed
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product LineeBioscience
Product TypePropidium Iodide
Unit SizeEach

Frequently asked questions (FAQs)

What is the concentration of eBioscience Propidium Iodide?

eBioscience Propidium Iodide is supplied at a concentration of 0.02 mg/mL and we recommend using 10 µL in a total staining volume of 200 µL.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.