DDM (n-dodecyl β-D-maltoside) (10%) - FAQs
DDM (n-dodecyl β-D-maltoside) (10%) - FAQs
View additional product information for DDM (n-dodecyl β-D-maltoside) (10%) - FAQs (BN2005)
1 product FAQs found
Can I use the NativePAGE Sample and Running buffers with NuPAGE Tris-Acetate gels for native applications?
NativePAGE Sample Buffers and Running buffers were developed specifically for use with the NativePAGE Bis-Tris gels. We do not recommend using these buffers for native applications with any other gels, including NuPAGE Tris-Acetate gels. When running Tris-Acetate gels under native conditions, we recommend using the Invitrogen Tris-Glycine Native Sample and running buffers.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.