MacConkey Broth Purple
MacConkey Broth Purple
Thermo Scientific™

MacConkey Broth Purple

Selectively detect and enumerate lactose‐fermenting Gram-negative bacteria in samples by observing a pH‐dependent colour change.
Have Questions?
Catalog NumberFill Weight
BO0376M100 to 101.5 g
Catalog number BO0376M
Price (USD)
-
Request A Quote
Fill Weight:
100 to 101.5 g
Selectively detect and enumerate lactose‐fermenting Gram-negative bacteria in samples by observing a pH‐dependent colour change using MacConkey Broth Purple.
Specifications
ColorPurple
Fill Weight100 to 101.5 g
Format125 mL Sirop Bottle
Packaging Type10 Sirop Bottles with screw cap closures in a box
Quantity10/Pk.
Shelf Life52 Weeks
Manufacturing OriginUnited Kingdom
Product TypeMicrobiology Media
Unit SizeEach
Contents & Storage
2°C to 25°C