Cold Filterable Vegetable Peptone Broth in BPCs
Thermo Scientific™

Cold Filterable Vegetable Peptone Broth in BPCs

Minimize prep time and maximize production for media fill trials with this ready-to-use peptone broth, an alternative to TSB, in BioProcess Containers.
Have Questions?
變更視圖buttonViewtableView
產品號碼QuantityFormat
BP0104C10 LBioprocess Container
BP0104A1 LBioprocess Container
BP0104E20 LBioprocess Container
產品號碼 BP0104C
價格 (HKD)
-
Quantity:
10 L
Format:
Bioprocess Container
Minimize prep time and maximize production for media fill trials with ready-to-use Thermo Scientific™ Cold Filterable Vegetable Peptone Broth in BPCs, a vegetable based alternative to TSB in BioProcess Containers. Simply connect the BPCs into your existing workflow to maximize process simulations and minimize secondary contamination.

Effective

  • Achieve effective process simulations, with minimized risk of secondary contamination
  • Just plug and play
  • Minimize preparation time
  • Satisfy regulatory compliance
  • Save time, reduce labour costs

Easy-to-Use

  • Innovative bag design and connector options for easy process integration
  • Standard, ready-to-use, off-the-shelf product range
  • No mixing, no sterilization, long shelf life at ambient storage
  • No Mycoplasma contamination
  • Qualified microbial recovery performance
  • Comprehensive validation package for media, bag/components and filling process
  • No filter blockages
  • No mixing required
  • No sterilization
  • No WFI (water for injection) required
  • No vessel cleaning

Pharmacopoeial-compliant Tryptone Soya Broth and Animal Derived Component Free (ADCF) vegetable peptone-based formulations are available.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

SmartSite™ is a trademark or registered trademark of the Cardinal Healthcare Inc.

ReadyMate™ is a trademark or registered trademark of the gE Healthcare BioSciences AB.

規格
ColorClear, straw
DescriptionCold Filterable Vegetable Peptone Broth in BPCs
FormatBioprocess Container
IncludesIncludes the following connectors: SmartSite, needle-free valve additional sampling port, Quick connect, +ö body (MPC), 1+ö triclamp and ReadyMate aseptic connector
Packaging Type1/Pk.
Quantity10 L
Shelf Life1 Year
Product TypeMicrobiology Media
Unit SizeEach
內容物與存放
2 to 25°C