Autosampler Enclosure
Thermo Scientific™
Autosampler Enclosure
Maintain ICP-OES and ICP-MS sample integrity with enclosures for the ESI SC-2DX and SC-4DX autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
BRE0008767Autosampler EnclosureESI SC-4 DX Autosampler
BRE0009103Autosampler EnclosureESI SC-2 DX Autosampler
Catalog number BRE0008767
Price (MXN)
-
Type:
Autosampler Enclosure
For Use With (Equipment):
ESI SC-4 DX Autosampler
Protect your samples and standards from airborne contamination with an enclosure designed for your ESI DX Autosampler. An exhaust port (50 mm, included) may be connected to the laboratory ventilation system to exhaust acid fumes and protect the laboratory environment and equipment.
Benefits of the ESI DX Autosampler Enclosures
  • Protects samples from airborne contamination
  • Exhaust acid fumes to protect the laboratory environment
  • Clear acrylic construction for easy sample viewing
  • Covers only the sample area—the electronics are kept outside the enclosure
  • Large hinged door provides easy access to sample racks at any time
  • Exhaust port (50 mm, included) connects to laboratory ventilation system
Compatible with ESI SC-2DX and SC-4DX Autosamplers
Specifications
For Use With (Equipment)ESI SC-4 DX Autosampler
TypeAutosampler Enclosure
Unit SizeEach