iSC-65 Autosampler Sample Tray
Thermo Scientific™
iSC-65 Autosampler Sample Tray
Optimize your laboratory workflow with the Thermo Scientific™ iSC-65 Autosampler Sample Tray with its versatile and efficient configuration.
Have Questions?
Catalog NumberType
BRE0032854Autosampler Sample Tray
Catalog number BRE0032854
Price (USD)
133.00
Each
Type:
Autosampler Sample Tray
Price (USD)
133.00
Each
This tray features a 10-position standard rack and can hold up to 4 sample vial racks, providing ample space for diverse sampling needs. With an integrated spill drain feature, it ensures hazardous substances are safely directed away, protecting personnel and equipment. Compatible with the iSC-65 Autosampler system, it guarantees reliable performance and ease of use.
The iSC-65 Autosampler Sample Tray has the following features:
  • 10-Position Standard Rack: Allows processing of multiple samples seamlessly.
  • Capacity for 4 Sample Vial Racks: Provides ample space for diverse sampling needs.
  • Integrated Spill Drain Feature: Ensures hazardous substances are safely directed away, protecting personnel and equipment.
  • Compatibility: Specifically designed for use with the iSC-65 Autosampler system.
Specifications
For Use With (Equipment)iSC-65 Autosampler
TypeAutosampler Sample Tray
Unit SizeEach