iSC-65 Spill Tray Drain Tubing
Thermo Scientific™
iSC-65 Spill Tray Drain Tubing
Effectively manage spills and minimize contamination risks, ensuring a safer and more efficient laboratory environment with Thermo Scientific™ iSC-65 Spill Tray Drain Tubing.
Have Questions?
Catalog NumberType
BRE0034191Spill Tray Drain Tubing
Catalog number BRE0034191
Price (USD)
30.20
Each
Type:
Spill Tray Drain Tubing
Price (USD)
30.20
Each
Designed for seamless integration with the iSC-65 Autosampler system, this high-quality tubing effectively channels spills away from work areas, minimizing the risk of contamination and exposure to hazardous substances. Easy to install and maintain, it enhances both safety and productivity in your lab. Invest in reliable performance and peace of mind with this essential component for spill management and containment.
Enhanced Safety
  • Protects personnel from exposure to hazardous substances
  • Reduces risk of contamination in the laboratory environment
  • Channels spills away from critical work areas
Specifications
MaterialTygon
For Use With (Equipment)iSC-65 Autosampler
TypeSpill Tray Drain Tubing
Unit SizeEach