Do you offer SSTR2 antibodies for western blot detection?
We offer 5 SSTR2 polyclonal antibodies that target human and have been verified for western blot detection (https://www.thermofisher.com/antibody/primary/target/sstr2/filter/proteintarget/SSTR2/application/Western%20Blot/species/Human). Their tested dilutions for western blot detection are listed below.
Catalog Number and related western blot dilution information:
PA3-109 1:250
711091 1-2 µg/mL
PA3-16736 1:100-1:1000
PA5-85431 1:500-1:3000
BS-1138R Assay Dependent
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Catalog Number and related western blot dilution information:
PA3-109 1:250
711091 1-2 µg/mL
PA3-16736 1:100-1:1000
PA5-85431 1:500-1:3000
BS-1138R Assay Dependent
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.