Regulatory requirements vary by country; products may not be available in your geographic area.
Priocheck™ Ruminant BVD/BD p80 Milk ELISA Kit
Thermo Scientific™

Priocheck™ Ruminant BVD/BD p80 Milk ELISA Kit

Single-well blocking ELISA kits enable sensitive and specific detection of pathogen-specific antibodies in animal samples, allowing for reliable and rapidRead more
Have Questions?
Catalog NumberQuantity
BVDAL55 x 96 well plates
Catalog number BVDAL5
Price (USD)
-
Quantity:
5 x 96 well plates
Single-well blocking ELISA kits enable sensitive and specific detection of pathogen-specific antibodies in animal samples, allowing for reliable and rapid screening and detection of seropositive animals.

How the kit works:

1. Samples are added to the specific antigen-coated microplate. If present in the sample, specific anti-pathogen antibodies bind to the antigen and form antigen-antibody complexes.
2 . After washing, an anti-pathogen peroxidase-labeled conjugate is added, which binds to any remaining unbound antigen.
3 . After a second washing, a chromogenic substrate is added. Color development results from oxidation of the substrate by the conjugated peroxidase.
4 . After stopping the reaction, results are read using an ELISA plate reader. Positive samples will show relatively weak or no color.

For veterinary use only.

Regulatory requirements vary by country; products may not be available in your geographic area.

Biological hazard safety

WARNING! BIOHAZARD. Biological samples such as tissues, body fluids, infectious agents, and blood of humans and other animals have the potential to transmit infectious diseases. All work should be conducted in properly equipped facilities using the appropriate safety equipment (for example, physical containment devices). Safety equipment also may include items for personal protection, such as gloves, coats, gowns, shoe covers, boots, respirators, face shields, safety glasses, or goggles. Individuals should be trained according to applicable regulatory and company/institution requirements before working with potentially biohazardous materials. Follow all applicable local, state/provincial, and/or national regulations. The following references provide general guidelines when handling biological samples in laboratory environment.

U.S. Department of Health and Human Services, Biosafety in Microbiological and Biomedical Laboratories (BMBL), 5th Edition, HHS Publication No. (CDC) 21-1112, Revised December 2009

World Health Organization, Laboratory Biosafety Manual, 3rd Edition, WHO/CDS/CSR/LYO/2004.11
Specifications
For Use With (Application)ELISA
For Use With (Equipment)ELISA Plate Reader
Product LinePriocheck
Quantity5 x 96 well plates
Target Organism ClassBorder Disease Virus (BDV), Bovine Viral Diarrhea Virus (BVDV)
Detection MethodColorimetric
Sample TypeMilk
Target SpeciesBovine, Caprine, Ovine, All Sensitive Species
Unit SizeEach