Find out about our new and improved Large Vessel Endothelial Supplement (LVES).
Human Umbilical Vein Endothelial Cells (HUVEC)
Gibco™
Human Umbilical Vein Endothelial Cells (HUVEC)
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from human umbilical vein and cryopreserved at the end of theRead more
Have Questions?
Catalog NumberQuantity
C0035C1 vial
Catalog number C0035C
Price (USD)
634.00
Each
Quantity:
1 vial
Price (USD)
634.00
Each
Primary Human Umbilical Vein Endothelial Cells (HUVEC) are isolated from human umbilical vein and cryopreserved at the end of the primary culture.
The cells are:
• Performance tested—at least 16 population doublings when cultured in Medium 200 supplemented with Large Vessel Endothelial Supplement (LVES)
• Guaranteed to be ≥ 70% viable
• Tested for mycoplasma, bacteria, yeast or other fungi, hepatitis B, hepatitis C, and HIV-1 viruses
Each vial contains over 0.5 x106 viable cells that have been cryopreserved at the end of the primary culture stage in a medium containing 10% DMSO.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Donor SourceSingle Donor
No. of Cells≥500000
Product LineCascade Biologics
Product TypeHuman Umbilical Vein Endothelial Cells
Quantity1 vial
Tested ForHepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
AgeNewborn (14 days or less)
Cell TypeEndothelial Cells (Umbilical Vein)
Culture TypeAdherent Cell Culture
FormCryopreserved
SpeciesHuman
Unit SizeEach
Contents & Storage
Store in liquid nitrogen.
Frequently asked questions (FAQs)
What types of large vessel endothelial cells do you offer?
We carry primary human umbilical vein endothelial cells (HUVEC) from one donor or pooled from multiple donors.
What media do you use during the first passage of primary cells?
We use various media and supplements for the various primary cells. Details can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-culture/primary-cell-culture.html).
How is the population doubling calculated for primary cells?
Cumulative population doublings are calculated from the seeding density and harvesting density from each culture level.
What are typical primary cell yields from tissue?
It varies a lot depending on the age of donor, size of tissues, locations, and conditions arrived to our facility. Sometime a small amount of tissue gives us many cells and sometimes a large amount of tissue gives us a very small lot. Typically, we can get 30-300 vials.
Are primary cells isolated from tissue using enzymatic digestion?
Yes, our primary cells are isolated from tissue using enzymatic digestion.
Citations & References (2)
Citations & References
Abstract
Generation of iPSC line epiHUVEC from human umbilical vein endothelial cells.
Authors:Matz P, Adjaye J,
Journal:
PubMed ID:26987922
Human umbilical vein endothelial cells (HUVECs) were used to generate the iPSC line epiHUVEC employing a combination of three episomal-based plasmids expressing OCT4, SOX2, NANOG, LIN28, c-MYC and KLF4. Pluripotency was confirmed both in vivo and in vitro. The transcriptome profile of epiHUVEC and the human embryonic stem cell line ... More
Angiogenic properties of dehydrated human amnion/chorion allografts: therapeutic potential for soft tissue repair and regeneration.
Authors:Koob TJ, Lim JJ, Massee M, Zabek N, Rennert R, Gurtner G, Li WW,
Journal:
PubMed ID:24817999
Chronic wounds are associated with a number of deficiencies in critical wound healing processes, including growth factor signaling and neovascularization. Human-derived placental tissues are rich in regenerative cytokines and have been shown in randomized clinical trials to be effective for healing chronic wounds. In this study, PURION® Processed (MiMedx Group, ... More