Can I purchase multiple batches of Human Epidermal Keratinocytes, adult (HEKa) (Cat. No. C0055C) from the same donor?
No, these cells are single donor lots so only one lot number is obtained from each donor.
Search
View additional product information for Human Epidermal Keratinocytes, adult (HEKa) - FAQs (C0055C)
1 product FAQs found