Product Image
Thermo Scientific™
Glass Thermometer for Lindberg/Blue M™ Shaking Circulating Water Baths
Use with Thermo Scientific™ SWB1122 and RSWB3222 Lindberg/Blue M water baths.
Have Questions?
Catalog NumberFor Use With (Equipment)
C01C-6Water Bath Models SWB1122, RSWB3222
Catalog number C01C-6
Price (MXN)
-
For Use With (Equipment):
Water Bath Models SWB1122, RSWB3222
The Thermo Scienfic™ Glass Thermometer is for use with Thermo Scientific™ SWB1122 and RSWB3222 Lindberg/Blue M water baths.
  • Temperature range: -10° to +110°C
  • With 1°C graduations
Specifications
DescriptionGlass Thermometer
For Use With (Equipment)Water Bath Models SWB1122, RSWB3222
Length (English)13 in.
Length (Metric)33 cm
TypeThermometer
Unit SizeEach