Gibco™

Human Epidermal Keratinocytes, adult, Animal Product-Free (HEKa-APF)

Catalog number: C0215C
Gibco™

Human Epidermal Keratinocytes, adult, Animal Product-Free (HEKa-APF)

Catalog number: C0215C
Catalog Number
C0215C
also known as C-021-5C
Unit Size
1 vial
Price (USD)
644.00
Your Price:
Availability
***
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C0215C
also known as C-021-5C
1 vial
644.00
Your Price:
***
Product Overview
Figures
Videos
Documents
FAQ
Citations & References
Additional Information
Recommendations
Recommendations
Primary human epidermal keratinoctyes from adult skin. Isolated, grown and cryopreserved at the end of the primary culture in an animal product-free environment.

• Viability at least 70%
• ≥500,000 viable cells⁄vial
• Capable of at least 25 population doublings
• Mycoplasma: Not Detected
• Hepatitis B: Not Detected
• Hepatitis C: Not Detected
• HIV-1: Not Detected
• Bacteria, yeast, and other fungi: Not Detected

Specifications

Age
Adult
Cell Type
Keratinocytes
Classification
Animal Origin-Free
Culture Type
Adherent Cell Culture
Donor Source
Single Donor
Form
Cryopreserved
Species
Human
Tested For
Hepatitis B (HBV), Hepatitis C (HCV), Sterility Testing (Bacteria & Fungi), Mycoplasma, Growth Performance Assay, HIV-1
Type
Human Epidermal Keratinocyte
Product Line
Cascade Biologics™

Contents & Storage

Liquid nitrogen temperature

Figures