Human Epidermal Melanocytes, neonatal, moderately pigmented donor, (HEMn-MP), Proliferating Culture, Prepared in Medium 254 and HMGS - FAQs
Human Epidermal Melanocytes, neonatal, moderately pigmented donor, (HEMn-MP), Proliferating Culture, Prepared in Medium 254 and HMGS - FAQs
View additional product information for Human Epidermal Melanocytes, neonatal, moderately pigmented donor, (HEMn-MP), Proliferating Culture, Prepared in Medium 254 and HMGS - FAQs (C10225PA)
1 product FAQs found
Do you have ethnicity information for your human epidermal melanocyte donor lines?
We do provide age/sex/pigmentation/ethnicity on the Certificate of Analysis (COA).