Human Epidermal Melanocytes, neonatal, moderately pigmented donor, (HEMn-MP), Proliferating Culture, Prepared in Medium 254 and HMGS-2

Catalog number: C10225PB

Gibco™  Related applications: Mammalian Cell Culture | Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Tertiary cultures established from cryopreserved cells, grown to approximately 50% confluence in T-25 flasks.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Age: Newborn (14 days or less)
Cell Type: Melanocytes
Culture Type: Adherent Cell Culture
Donor Attributes: Moderately Pigmented
Donor Source: Single Donor
Form: Proliferating Culture
Product Line: Cascade Biologics®
Product Size: 3 flasks
Species: Human
Tests Performed: Growth Performance Assay, HIV-1, Hepatitis B (HBV), Hepatitis C (HCV), Mycoplasma, Sterility Testing (Bacteria & Fungi)
Shipping Condition: Room Temperature

Contents & storage

Upon receipt, follow the instructions in the manual.

Documents

Manuals & protocols