Countess™ Cell Counting Chamber Slides with Trypan Blue Stain, disposable, 1250 Slides, 100 mL bottle - FAQs
Countess™ Cell Counting Chamber Slides with Trypan Blue Stain, disposable, 1250 Slides, 100 mL bottle - FAQs
View additional product information for Countess™ Cell Counting Chamber Slides - FAQs (A40008469, C10283, C10315, C10314, A25750, C10228, C10313, C10312)
1 product FAQs found
Do you offer an alternative for Countess Cell Counting Chamber Slides (Cat. no. C10283) for Countess 3 and Countess 3 FL Automated Cell Counter?
Yes. The alternative options for Countess Cell Counting Chamber Slides (Cat. No. C10283) and for Countess 3 and Countess 3 FL Automated Cell Counter can be found using the link below:
Countess Cell Counting Chamber Slides (Cat. Nos. C10228, C10312, C10313, C10314, C10315)