Countess™ Cell Counting Chamber Slides with Trypan Blue Stain, disposable, 5000 Slides, 2 x 100 mL bottles - FAQs
Countess™ Cell Counting Chamber Slides with Trypan Blue Stain, disposable, 5000 Slides, 2 x 100 mL bottles - FAQs
View additional product information for Countess™ Cell Counting Chamber Slides - FAQs (A40008469, C10283, C10315, C10314, A25750, C10228, C10313, C10312)
2 product FAQs found
Do you offer an alternative for Countess Cell Counting Chamber Slides (Cat. no. C10283) for Countess 3 and Countess 3 FL Automated Cell Counter?
Yes. The alternative options for Countess Cell Counting Chamber Slides (Cat. No. C10283) and for Countess 3 and Countess 3 FL Automated Cell Counter can be found using the link below:
Countess Cell Counting Chamber Slides (Cat. Nos. C10228, C10312, C10313, C10314, C10315)
What Countess Automated Cell Counter instruments are compatible with the Countess Cell Counting Chamber Slides (Cat. No. C10228, C10312, C10314, C10315)?
The Countess Cell Counting Chamber Slides (Cat. No. C10228, C10312, C10314, C10315) are compatible with all Countess Automated Cell Counter instruments.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center .