Click-iT™ Plus OPP Alexa Fluor™ 594 Protein Synthesis Assay Kit - FAQs
Click-iT™ Plus OPP Alexa Fluor™ 594 Protein Synthesis Assay Kit - FAQs
View additional product information for Click-iT™ Plus OPP Alexa Fluor™ 594 Protein Synthesis Assay Kit - FAQs (C10457)
1 product FAQs found
I will be performing a cell proliferation assay using Click-iT EdU kit. At what point can I stop overnight, or do I have to perform all the steps continuously?
One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.
Find additional tips, troubleshooting help, and resources within our Cell Viability, Proliferation, Cryopreservation, and Apoptosis Support Center.